Learn More
Waters Corp Waters Column, C18, 90Å, 2.7 μm, 3 mm × 50 mm
Supplier:  Waters Corp 186007370
Encompass_Preferred
CORTECS C18 Columns are general purpose, high-efficiency columns based on a solid-core particle. They offer balanced retention of acids, bases, and neutrals at low and mid-range pH, providing excellent efficiency, resolution, and retention for complex analyte mixtures. These columns are designed to improve performance by lowering the three terms of the van Deemter equation: the A term, the B term, and the C term, making them ideal lab equipment to resolve complex mixtures or maintain resolution while increasing throughput.
  • General purpose, high-efficiency columns.
  • Based on a solid-core particle.
  • Offers balanced retention of acids, bases, and neutrals at low and mid-range pH.
  • Provides excellent efficiency, resolution, and retention for complex analyte mixtures.
  • Improves performance by lowering each of the three terms of the van Deemter equation.
Catalog No. NC1795575