Learn More
Waters Corp CORTECS C18 Column, 90Å, 1.6 μm, 3 mm x 100 mm
Supplier:  Waters Corp 186007100
Encompass_Preferred
CORTECS C18 Columns are high-efficiency columns with solid-core particles, offering balanced retention for acids, bases, and neutrals across a 2 to 8 pH range. Designed for versatility, they provide excellent peak shapes and enhanced performance by reducing axial diffusion and improving heat transport, leading to increased sample throughput and superior peak capacities.
  • Solid-core particle technology ensures high efficiency and performance
  • Provides balanced retention for acids, bases, and neutrals (pH 2 to 8)
  • Delivers excellent peak shapes for basic analytes at low pH
  • Maximizes sample throughput and offers superior peak capacities
  • Achieves sharper, narrower peaks compared to fully porous particles
  • Operates effectively up to 45 °C
  • Best for increased resolution, speed, and sensitivity
  • Compatible with pre-columns for extended column life and consistent performance
Catalog No. 50-239-1584