Learn More
Waters Corp Column, C18, 2.7 μm, 90 Å, 4.6 mm × 100 mm
Supplier:  Waters Corp 176003315
Encompass_Preferred
These high-efficiency solid-phase columns offer balanced retention of acids, bases, and neutrals across a wide pH range. This specific column has a length of 100 mm, an inner diameter of 4.6 mm, and a particle size of 2.7 μm, designed for high-speed applications.
  • High-efficiency columns based on a solid-core particle for general purpose use.
  • Offer balanced retention of acids, bases, and neutrals across a wide pH range.
  • Provide good efficiency, resolution, and retention for complex analyte combinations.
  • Improve performance by lowering van deemter equation terms.
  • Manufactured to high industry standards (ISO 9002, ISO 13485, cGMP).
  • Ligand provides good retention and stability.
  • Increase throughput and resolution with faster separations at low operating back pressures.
Catalog No. 50-239-2869