Learn More
Waters Corp Column, CORTECS C18, 90Å, 2.7 μm, 3 mm x 75 mm
Supplier:  Waters Corp 186007371
Encompass_Preferred
CORTECS C18 Columns are general purpose, high-efficiency columns based on a solid-core particle that offer balanced retention of acids, bases & neutrals at low and mid-range pH. These columns allow for enhanced sample throughput and can be used to resolve complicated combinations or retain resolution while boosting throughput.
  • Offers balanced retention of acids, bases & neutrals at low and mid-range pH.
  • Solid-core particles decrease the three terms in the van Deemter equation (A, B, and C terms) for improved performance.
  • Enables enhanced sample throughput and allows the use of shorter columns for comparable peak capacities and faster re-equilibration.
  • Helps to run more samples in the same amount of time without sacrificing separation performance, saving lab operation costs.
  • Certificates of analysis (COAs) are available online at www.waters.com/coa.
Catalog No. 50-239-1691