Learn More
Waters Corp C18+ Column, 2.7 μm, 3 mm × 100 mm
Supplier:  Waters Corp 176003297
Encompass_Preferred
The CORTECS C18+ Column is a high-efficiency, general-purpose reversed-phase column designed to deliver excellent peak shape for basic analytes at low pH. This column features a solid-core particle technology that boosts performance and improves throughput.
  • Delivers excellent peak shape for basic analytes at low pH.
  • Solid-core particles enhance performance and improve heat transfer.
  • Allows for faster analysis speed without sacrificing separation performance.
  • Enables increased sample throughput with equivalent or superior peak capacities.
  • C18 ligand retains and stabilizes data effectively.
  • Withstands pressures up to 18,000 psi (1240 Bar) and temperatures up to 45 °C.
  • Compatible with VanGuard Cartridges to extend analytical column lifetime.
Catalog No. 50-239-0451