Learn More
Waters Corp CORTECS C18+ Column, 90 Å Pore Size, 2.7 μm Particle Size, 2.1 mm ID x 100 mm L
Supplier:  Waters Corp 186007397
Encompass_Preferred
These columns are general purpose, charged surface, high efficiency C18 columns based on solid-core particles, designed to deliver excellent peak shape for basic analytes at low pH. They enable resolution of complex mixtures and increased throughput by lowering the A, B, and C terms of the van Deemter equation.
  • Delivers excellent peak shape for basic analytes at low pH.
  • Solid-core particle technology enhances performance by reducing axial diffusion and improving heat transfer.
  • Increases sample throughput while maintaining or improving peak capacities by raising flow rates.
  • Allows shorter column lengths for faster separations without compromising resolution.
  • Offers outstanding selectivity for resolution.
  • Innovative particle-surface modifications enable faster method development.
  • Provides excellent chromatographic performance across a wide pH range of 2 to 8.
Catalog No. 50-203-7987