Learn More
Waters Corp Column, C8, 1.6 μm Particle, 2.1 mm ID, 75 mm Length
Supplier: Waters Corp 186008400
These columns are designed for general purpose method development under low pH test conditions. The C8 ligand is preferred for some assays due to its less hydrophobic nature, making it less retentive compared to C18 phases.
- C8 chemistry for reversed phase separation
- Silica particle substrate
- pH range of 2 to 8
- Temperature limits up to 45°C
- Maximum pressure of 18000 psi (1240 bar)
- Endcapped
- Low silanol activity
- Spherical particle shape
- 1.6 μm particle size
- Parker-style endfitting
- 90 Å pore size
- Surface area: 100
- Compatible with UPLC and UHPLC systems
- Solid core particle technology
- USP classification: L7
- 2.1 mm inner diameter
- 75 mm length
- 4.5% carbon load
- eCord enabled
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.