Learn More
Waters Corp CORTECS C8 Column, 90Å, 1.6 μm, 3 mm X 75 mm
Supplier: Waters Corp 176003836
CORTECS C8 columns are used for general purpose method development under low pH test conditions. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to C18 phases.
- Chemistry: C8, reversed phase separation mode
- Particle substrate: Silica, spherical particle shape, solid core technology
- pH range: 2 to 8 pH
- Temperature limit: 45°C
- Maximum pressure: 18000 psi (1240 Bar)
- Particle size: 1.6 μm
- Pore size: 90 Å
- Inner diameter: 3 mm
- Length: 75 mm
- Carbon load: 4.5%
- Endcapped: Yes, with low silanol activity
- Endfitting type: Parker-style
- System compatibility: UPLC, UHPLC
- USP classification: L7
- eCord enabled: Yes
- Units per package: 3
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.