Learn More
Waters Corp CORTECS C8 Column, 90Å, 2.7 μm, 4.6 mm X 100 mm
Supplier: Waters Corp 176003817
These C8 columns are designed for general purpose method development in low pH test conditions. The C8 ligand is less hydrophobic and less retentive compared to C18 phases, making them suitable for specific assays.
- Designed for general purpose method development in low pH test conditions.
- Less hydrophobic and less retentive C8 ligand compared to C18 phases.
- Chemistry: C8.
- Particle substrate: Silica.
- pH range: 2 to 8 pH.
- Temperature limits: 45°C.
- Maximum pressure: 18000 psi (1240 bar).
- Particle size: 2.7 μm.
- Pore size: 90 Å.
- Inner diameter: 4.6 mm.
- Length: 100 mm.
- Units per package: 3.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.