Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Column, CORTECS C8, 90Å, 2.7 μm, 4.6 × 75 mm

Supplier:  Waters Corp 186008370

Encompass_Preferred

These CORTECS 2.7 μm columns, featuring solid-core particle technology, reduce operational backpressure and enhance resolution. They significantly improve throughput and efficiency across both HPLC and UHPLC systems.

  • Reduces operational backpressure by 25% compared to fully porous packing materials of similar particle sizes.
  • Enables increased resolution using longer columns.
  • Improves throughput by up to 5-fold versus conventional 5 μm HPLC columns.
  • Provides more efficient separations than traditional 3.5 μm and 5 μm packing materials.
  • Compatible with both HPLC and UHPLC instrumentation, maximizing asset utilization.
  • Utilizes solid-core particle technology for optimized separation performance.
  • Supported by VanGuard Cartridge Columns for extended lifetime and protection.

Catalog No. 50-239-2004


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.