Learn More
Waters Corp HPLC Column, C8, 50 mm, 4.6 mm, 2.7 μm
Supplier:  Waters Corp 176003815
Encompass_Preferred
CORTECS C8 columns are suitable for general method development in low pH conditions. This 50 mm long column has a 4.6 mm inner diameter and 2.7 μm particle size. Featuring C8 ligand for less hydrophobicity than C18, CORTECS columns offer diverse materials and innovative surface modifications, allowing faster separations and higher resolution than fully porous particle columns.
  • Optimized for low pH method development.
  • C8 ligand provides reduced hydrophobicity and retention versus C18 phases.
  • Available in reversed-phase and HILIC configurations.
  • Utilizes diverse column materials and innovative surface modifications.
  • Achieves faster separations and higher resolution than fully porous particle columns.
Catalog No. 50-239-0495