Learn More
Waters Corp CORTECS UPLC HILIC Column, 1.6 μm, 90 Å, 3.0 x 30 mm
Supplier:  Waters Corp 176003161
Encompass_Preferred
The CORTECS UPLC HILIC Column, with 1.6 μm particle size and 90 Å pore size, is designed for high-efficiency separations. It provides excellent resolution, speed, and sensitivity for a wide array of compounds.
  • Provide the highest column efficiencies when used with low-dispersive UPLC instrumentation.
  • Produce sharper, narrower peaks compared to fully porous particles of similar size.
  • Recommended for increased resolution, speed, and sensitivity.
  • HILIC chemistry offers flexibility to rapidly separate a wide array of compounds.
  • Available in seven unique CORTECS chemistries, including HILIC.
Catalog No. 50-239-2480