Learn More
Waters Corp CORTECS HILIC Column, 1.6 μm, 90 Å, 2.1 mm x 100 mm
Supplier:  Waters Corp 186007106
Encompass_Preferred
CORTECS HILIC Columns are high-efficiency columns using solid-core particles, designed for retaining extremely polar analytes and offering orthogonal selectivity to C18 columns. They provide faster separations and higher throughput through Hydrophilic Interaction Chromatography (HILIC) mode.
  • High-efficiency columns with solid-core particles.
  • Retains extremely polar analytes.
  • Offers orthogonal selectivity to C18 columns.
  • Provides faster separations for greater throughput.
  • Uses HILIC mode for polar compound retention.
  • Compatible with high organic solvent mobile phases, improving MS response.
Catalog No. 50-239-2354