Learn More
Waters Corp CORTECS HILIC Column, 90Å, 2.7 μm, 2.1 mm x 50 mm
Supplier:  Waters Corp 186007380
Encompass_Preferred
This column is a CORTECS HILIC column designed for the retention of very polar compounds using hydrophilic-interaction chromatography (HILIC). It offers an orthogonal separation mode to reversed-phase for mixtures of ionizable and very polar compounds, leveraging solid-core particle technology.
  • Enables seamless method transfer between UPLC, UHPLC, and HPLC platforms.
  • Exhibits maximum chromatographic performance.
  • Retains very polar bases, while acidic, neutral, and non-polar compounds will have limited retention.
  • Combination of acetonitrile-rich mobile phase and polar, unbonded stationary phase provides increased mass spectrometry response.
  • Allows direct injection of organic extracts from common sample preparation techniques such as solid-phase extraction, liquid-liquid extraction, and protein precipitation.
Catalog No. 50-239-1407