Learn More
Waters Corp CORTECS UPLC HILIC Column, 90Å Pore, 2.7 μm Particle, 2.1 × 150 mm
Supplier:  Waters Corp 176003283
Encompass_Preferred
The CORTECS UPLC HILIC Column is designed for high-performance liquid chromatography, offering superior efficiency and sharper peaks. It is ideal for increased resolution, speed, and sensitivity in separations, particularly when used with low-dispersive UPLC instrumentation.
  • Solid-core particle type.
  • L3 USP class number.
  • pH range of 1 to 5.
  • Temperature limits: 45 °C for both low and high pH.
  • Surface area of 100 m2/g.
  • Performance standard is HILIC QC Reference Material.
  • Provides highest column efficiencies when used with low-dispersive UPLC instrumentation.
  • Produces sharper, narrower peaks compared to fully porous particles of similar size.
  • Suitable for increased resolution, speed, and sensitivity in separations.
Catalog No. 50-239-2051