Learn More
Waters Corp HPLC Column, 2.7 μm, 90 Å, 2.1 mm × 50 mm, 3 pack
Supplier:  Waters Corp 176003280
Encompass_Preferred
These HILIC columns are designed for the retention of highly polar compounds using hydrophilic-interaction chromatography (HILIC), offering an orthogonal separation mode compared to reversed-phase chromatography for mixtures of ionizable and very polar compounds.
  • Utilizes solid-core particles for maximized efficiency, throughput, and performance.
  • Features a polar, unbonded HILIC stationary phase.
  • Offers excellent efficiency, batch-to-batch reproducibility, and orthogonal selectivity for extremely polar analytes.
  • Ensures enhanced resolution and peak capacity.
  • Provides a 25% reduction in operating backpressure, increasing throughput.
  • Resists bonded phase loss for long column lifetimes.
Catalog No. 50-239-0814