Learn More
Waters Corp Column, Phenyl, 90Å, 2.7 μm, 3 mm x 75 mm
Supplier:  Waters Corp 176003796
Encompass_Preferred
These columns offer complementary selectivity to C18 stationary phases. Their particle morphology delivers high efficiency and reduced backpressures compared to fully porous particles of similar size. They ensure ultra-low column MS bleed, extended lifetimes, and superior peak shape.
  • High efficiency
  • Lower backpressures
  • Ultra-low column MS bleed
  • Long lifetimes
  • Excellent peak shape
Catalog No. 50-239-2665