Learn More
Waters Corp CORTECS Phenyl Column, 90Å, 2.7 μm, 4.6 mm x 30 mm
Supplier:  Waters Corp 176003799
Encompass_Preferred
CORTECS Phenyl columns offer complementary selectivity compared to C18 stationary phases. Their particle morphology provides high efficiency and lower backpressures than fully porous particles of similar size. CORTECS Phenyl also features ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Complementary selectivity to C18 stationary phases
  • High efficiency
  • Lower backpressures compared to fully porous particles of similar particle size
  • Ultra-low column MS bleed
  • Long lifetimes
  • Excellent peak shape
Catalog No. 50-239-2816