Learn More
Waters Corp Waters Corporation Column, C18+, 1.6 μm, 2.1 mm x 100 mm
Supplier:  Waters Corp 186010428
Encompass_Preferred
The CORTECS Premier C18+ Column is a high-efficiency C18 column featuring solid-core particle technology, optimized for excellent peak shape with basic analytes at low pH. It integrates MaxPeak High Performance Surface hardware to minimize analyte/surface interactions, enhancing recovery, sensitivity, and data reproducibility.
  • Delivers excellent peak shape for basic analytes at low pH.
  • Reduces unwanted analyte/surface interactions using MaxPeak High Performance Surface hardware.
  • Improves sensitivity and peak shapes.
  • Enhances analyte recovery.
  • Allows for simpler mobile phases by reducing the need for complex additives.
  • Provides maximum efficiency with solid-core technology.
  • Increases sample throughput.
  • Lowers column backpressure compared to fully porous particles of equivalent size.
Catalog No. NC2294336