Learn More
Waters Corp CORTECS Premier C18 Column, 2.7 μm, 2.1 mm x 50 mm
Supplier:  Waters Corp 186010441
Encompass_Preferred
CORTECS Premier Columns combine solid-core particle technology with MaxPeak High Performance Surfaces (HPS) for superior chromatographic performance. They are designed for precision, delivering exceptional peak shapes, low backpressure, and reliable data across a wide range of analytes.
  • Solid-core particles provide higher resolution and faster throughput.
  • MaxPeak HPS technology improves data quality and peak shape for metal-sensitive analytes.
  • Versatile for small molecule and biomolecule LC-MS applications.
  • Scalable performance enables seamless method transfer across LC systems.
  • Low backpressure allows high-speed separations.
  • Delivers consistent results from the first injection.
  • Achieve up to 3x more throughput with shorter run times.
  • Enhanced signal and accuracy for low-abundance or reactive compounds.
Catalog No. NC2294353