Learn More
Waters Corp CORTECS Columns, Premier C18, 2.7 um, 2.1 mm x 50 mm
Supplier:  Waters Corp 186010444
Encompass_Preferred
These general purpose, high-efficiency columns are based on a solid-core particle, offering balanced retention of acids, bases and neutrals at low and mid-range pH. MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions, preventing poor peak shape and signal loss. The VanGuard Fit design protects the column from contaminants, extending its lifetime.
  • General purpose, high-efficiency columns
  • Based on a solid-core particle
  • Offer balanced retention of acids, bases and neutrals at low and mid-range pH
  • Utilize MaxPeak High Performance Surface hardware technology
  • Significantly reduces unwanted analyte/surface interactions that can lead to poor peak shape
  • Reduces losses in signal intensity
  • VanGuard Fit column design protects the column from contaminants
  • Increases column lifetime
Catalog No. 50-239-1976