Learn More
Waters Corp CORTECS Premier C18 VanGuard FIT Column, 2.7 μm, 2.1 mm, 150 mm
Supplier:  Waters Corp 186010446
Encompass_Preferred
These general-purpose, high-efficiency columns are based on a solid-core particle, offering balanced retention for acids, bases, and neutrals at low and mid-range pH. They incorporate MaxPeak High Performance Surface hardware technology, which significantly reduces unwanted analyte/surface interactions that can lead to poor peak shape and losses in signal intensity. The column design protects the column from contaminants and increases column lifetime.
  • MaxPeak High Performance Surface hardware technology.
  • VanGuard Fit column design.
  • Solid-core particle.
Catalog No. 50-239-1986