Learn More
Waters Corp CORTECS Premier C18+ VanGuard FIT Column, 2.7 μm, 4.6 x 50 mm
Supplier:  Waters Corp 186010468
Encompass_Preferred
The CORTECS Premier FIT Column is designed to enhance chromatographic separations and extend column lifetime. It utilizes VanGuard FIT technology to protect the analytical column from particulate and chemical contamination, while MaxPeak High Performance Surfaces minimize undesirable analyte/surface interactions for improved sensitivity and recovery.
  • Protects columns from particulate and chemical contamination.
  • Ensures consistent chromatographic performance and stable backpressure.
  • Minimizes analyte/surface interactions for enhanced recovery and sensitivity.
  • Improves peak shapes and reduces column conditioning times.
  • Simplifies mobile phase requirements.
  • Contributes to faster method development and reliable data.
Catalog No. 50-239-2835