Learn More
Waters Corp Waters CORTECS Premier T3 Column, C18, 2.7 μm, 2.1 mm x 50 mm
Supplier:  Waters Corp 186010472
Encompass_Preferred
The CORTECS Premier T3 Column is designed for high-performance liquid chromatography, compatible with 100% aqueous mobile phases. It utilizes MaxPeak High Performance Surface hardware technology to reduce unwanted analyte/surface interactions, enhancing chromatographic performance.<ul><li>MaxPeak High Performance Surfaces technology improves analyte recovery and sensitivity.</li><li>Solid-core particle technology offers high efficiency and lower backpressure.</li><li>Compatible with 100% aqueous mobile phases.</li><li>Provides excellent retention for polar and non-polar analytes.</li><li>Features C18 chemistry with silica particle substrate.</li><li>Particle size of 2.7 μm.</li><li>Pore size of 120 Å.</li></ul>
Catalog No. NC2294373