Learn More
Waters Corp CORTECS Premier T3 VanGuard FIT Column, Reversed Phase, 2.7 μm, 2.1 x 100 mm
Supplier:  Waters Corp 186010476
Encompass_Preferred
The CORTECS Premier T3 VanGuard FIT Column is a high-performance liquid chromatography column for reversed-phase separations. It features a 2.7 μm silica particle with T3 bonding technology for robust retention of analytes. This column integrates MaxPeak High Performance Surface hardware to mitigate interactions and a VanGuard FIT design for extended lifetime.
  • Compatible with 100% aqueous mobile phases.
  • High efficiency and reduced backpressure.
  • Ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • MaxPeak High Performance Surface technology minimizes unwanted analyte interactions.
  • VanGuard FIT design protects against contaminants, removes particulate and chemical contamination, and extends column longevity.
  • Ensures consistent connections and easy installation with a quarter-turn.
  • Quickly restores performance by replacing the VanGuard FIT Cartridge.
Catalog No. 50-239-2590