Learn More
Waters Corp CORTECS UPLC Column, Shield RP18, 90Å, 2.7 μm, 3.0 mm x 50 mm
Supplier:  Waters Corp 176003918
Encompass_Preferred
The CORTECS UPLC Shield RP18 Column is designed for high-performance liquid chromatography. It features a Shield RP18 stationary phase with a 2.7 μm particle size and 90 Å pore size, offering efficient separations for various analytical applications.
  • Shield RP18 stationary phase for optimal separations.
  • 2.7 μm particle size ensures high resolution.
  • 90 Å pore size provides suitability for a range of analytes.
  • Column dimensions are 3.0 mm I.D. x 50 mm length.
  • Supplied in a convenient 3-pack.
Catalog No. 50-239-2923