Learn More
Waters Corp CORTECS Shield RP18 VanGuard Pre-column, 1.6 μm, 2.1 mm, 5 mm, 90 Å
Supplier:  Waters Corp 186008713
Encompass_Preferred
These pre-columns are designed to extend analytical column lifetime and performance by removing particulate and chemical contamination from the mobile phase. Optimized for analytical columns utilizing 1.6 μm sorbent particles, they are ideal for high-capacity biological fluid purification or temperature control applications.
  • Extends analytical column lifetime and performance.
  • Removes particulate and chemical contamination from mobile phase.
  • Optimized for analytical columns with 1.6 μm sorbent particles.
  • Manufactured in a cGMP, ISO 9001 certified plant.
  • Packing material chromatographically tested for repeatable performance.
Catalog No. 50-239-2274