Learn More
Waters Corp CORTECS Solid Core HILIC Column 90Å, 1.6 μm, 3 mm × 100 mm
Supplier:  Waters Corp 176003164
Encompass_Preferred
CORTECS HILIC Columns are high-efficiency columns based on a solid-core particle designed to retain extremely polar analytes, offering orthogonal selectivity compared to C18 columns.
  • High-efficiency columns
  • Designed to retain extremely polar analytes
  • Offers orthogonal selectivity versus C18 columns
Catalog No. 50-239-1209