Learn More
Waters Corp Column, Phenyl, 90Å Pore, 2.7 μm Particle
Supplier:  Waters Corp 186008332
Encompass_Preferred
The Waters Corporation CORTECS Solid Core Phenyl Column offers high efficiency and reduced backpressure, providing excellent resolution with standard LC instrumentation. It delivers alternative selectivity to C8 and C18 stationary phases, enhancing resolution for complex samples, and features ultra-low MS bleed, long lifetime, and good peak shape.
  • Provides complementary selectivity to C18 stationary phases.
  • Offers high efficiency and lower backpressures.
  • Ensures ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Produced in a cGMP, ISO 9001 accredited facility using ultra-pure reagents.
  • Each batch is chromatographically tested for consistent performance.
  • Column documents are available digitally for easy access.
Catalog No. 50-239-2517