Learn More
Waters Corp CORTECS Column, 120Å, 2.7 μm, 4.6 mm × 75 mm
Supplier:  Waters Corp 186008493
Encompass_Preferred
These columns are compatible with 100% aqueous mobile phases and offer excellent retention for both polar and non-polar analytes. The particle morphology provides high efficiency and lower backpressure compared to fully porous particles of similar size. They also deliver ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Compatible with 100% aqueous mobile phases
  • Excellent retention for polar and non-polar analytes
  • High efficiency and lower backpressure
  • Ultra-low column MS bleed
  • Long lifetimes
  • Excellent peak shape
  • Low Silanol Activity
  • Spherical Particle Shape
  • Solid Core Particle Technology
Catalog No. 50-239-2372