Learn More
Waters Corp WATERS CORPORATION CORTECS UPLC T3 Column, 120Å, 2.7 μm, 2.1 × 75 mm
Supplier:  Waters Corp 176003878
Encompass_Preferred
CORTECS UPLC T3 Columns, packed with 2.7 μm particles, are engineered for high-performance liquid chromatography applications. These columns deliver superior efficiency and produce sharper, narrower peaks, which contributes to enhanced resolution, speed, and sensitivity in analytical separations.
  • Engineered for high-performance liquid chromatography applications.
  • Packed with 2.7 μm particles.
  • Delivers superior efficiency.
  • Produces sharper, narrower peaks.
  • Enhances resolution, speed, and sensitivity in analytical separations.
  • Suitable for use with HPLC and UHPLC systems.
Catalog No. 50-239-2221