Learn More
Waters Corp CORTECS T3 Column, 2.7 μm, 120 Å, 2.1 mm x 75 mm
Supplier:  Waters Corp 186008483
Encompass_Preferred
CORTECS T3 columns are compatible with 100% aqueous mobile phases, offering excellent retention for both polar and non-polar analytes. Their particle morphology provides high efficiency and lower backpressure compared to fully porous particles of a similar size. These columns also deliver ultra-low column MS bleed, long lifetimes, and excellent peak shape, leveraging solid-core particle technology for increased resolution, speed, and sensitivity.
  • Compatible with 100% aqueous mobile phases.
  • Excellent retention for polar and non-polar analytes.
  • High efficiency.
  • Lower backpressure compared to fully porous particles.
  • Ultra-low column MS bleed.
  • Long lifetimes.
  • Excellent peak shape.
  • Utilizes solid-core particle technology.
  • Designed to produce sharper, narrower peaks.
  • Increased resolution, speed, and sensitivity.
Catalog No. 50-239-1478