Learn More
Waters Corp CORTECS T3 Column, 120 Å, 2.7 μm, 3 mm × 50 mm
Supplier:  Waters Corp 176003882
Encompass_Preferred
CORTECS T3 columns are compatible with 100% aqueous mobile phases, providing excellent retention for both polar and non-polar analytes. The particle morphology offers high efficiency and lower backpressure compared to fully porous particles of similar size. These columns deliver ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Compatible with 100% aqueous mobile phases
  • Provides excellent retention for polar and non-polar analytes
  • Offers high efficiency
  • Ensures lower backpressure compared to fully porous particles of similar size
  • Delivers ultra-low column MS bleed
  • Ensures long lifetimes
  • Provides excellent peak shape
  • Features T3 Bonding Technology for enhanced performance
Catalog No. 50-239-0742