Learn More
Waters Corp CORTECS Column, 90Å, 2.7 μm, 4.6 mm, 100 mm
Supplier:  Waters Corp 186008341
Encompass_Preferred
These phenyl columns offer complementary selectivity to C18 stationary phases. Their particle morphology delivers high efficiency and lower backpressures compared to fully porous particles of similar size. They also provide ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Offers complementary selectivity to C18 stationary phases.
  • Provides high efficiency and lower backpressures.
  • Ensures ultra-low column MS bleed.
  • Delivers long lifetimes and excellent peak shape.
  • Features spherical solid core silica particles.
  • Compatible with UPLC and UHPLC systems.
  • Designed for a pH range of 2 to 8.
  • Resistant to pressures up to 18000 psi (1240 Bar).
Catalog No. 50-138-4817