Learn More
Med Vet International Cortical Screw, J0416F, 2mm x 16mm, 6 Count
Supplier:  Med Vet International JOR416F
Encompass_Preferred
Available in 6mm-24mm lengths, the spherical head of the screw produces compression when used in conjunction with 2mm compression plate.
Catalog No. 50-005-82253