Learn More
Med Vet International Cortical Screw, J0416G, 2mm x 18mm, 6 Count
Supplier: Med Vet International JOR416G
Available in 6mm-24mm lengths, the spherical head of the screw produces compression when used in conjunction with 2mm compression plate.