Learn More
Med Vet International Cortical Screws, J0416H, 2mm x 20mm, 6 Count
Supplier: Med Vet International JOR416H
Available in 6mm-24mm lengths, the spherical head of the screw produces compression when used in conjunction with 2mm compression plate.