Learn More
Cayman Chemical CortIcostatInhumntrIflu 100ug
SDP
Supplier:  Cayman Chemical 24888100 UG
Encompass_Preferred
A corticostatic peptide; inhibits ACTH-stimulated corticosterone production in rat adrenal cells (IC50 = 0.7 μM); reduces mitogen-stimulated release of IFN-γ and IL-6 in human PBMCs
Catalog No. 50-293-4685