Learn More
Medchemexpress LLC Corticosterone (17-Deoxycortisol) | 50-22-6 | 99.8% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB1618500MG
Encompass_Preferred
Corticosterone (17-Deoxycortisol) is an orally active, adrenal cortex-produced glucocorticoid that plays a role in regulating neuronal functions of the limbic system. As an endogenous metabolite, it falls under the steroids structure classification.
  • Orally active glucocorticoid
  • Regulates neuronal functions of the limbic system
  • Increases Rab-mediated AMPAR membrane traffic
  • Interferes with dendritic cell maturation, showing an immunosuppressive effect
  • High purity of 99.77%
  • Supplied as a white to off-white solid
Catalog No. 50-004-11707