Learn More
Cayman Chemical Cortisol ELISA Kit, 96 solid wells
SDP
Supplier:  Cayman Chemical 50036096SOLIDWELLS
Encompass_Preferred
Cayman’s Cortisol ELISA is a competitive assay that provides accurate measurements of cortisol within the range of 6.6-4,000 pg/ml
Catalog No. NC9065281