Learn More
Cayman Chemical Cortisol ELISA Kit, 96 strip wells
SDP
Supplier:  Cayman Chemical 50036096STRIPWELLS
Encompass_Preferred
Cayman’s Cortisol ELISA is a competitive assay that provides accurate measurements of cortisol within the range of 6.6-4,000 pg/ml
Catalog No. NC1283721