Learn More
Medchemexpress LLC Cortisol sulfate sodium | 1852-36-4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN8460A10MG
Encompass_Preferred
Cortisol sulfate sodium, also known as Cortisol 21-sulfate, is a metabolite of Cortisol. It functions as a specific ligand for intracellular transcortin.
  • Metabolite of cortisol
  • Specific ligand for intracellular transcortin
  • Affects behavior through direct action on the central nervous system
Catalog No. 50-004-19237