Learn More
Medchemexpress LLC Cortisol sulfate sodium | 1852-36-4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN8460A100MG
Encompass_Preferred
Cortisol sulfate sodium (Cortisol 21-sulfate) is a metabolite of Cortisol and a specific ligand for intracellular transcortin. This product is intended for research use only and is not for sale to patients.
  • Specific ligand for intracellular transcortin
  • Metabolite of Cortisol
  • Solid appearance
  • White to off-white color
Catalog No. 50-004-19238