Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cortisol sulfate sodium | 1852-36-4 | 50 MG
SDP

Supplier:  Medchemexpress LLC HYN8460A50MG

Encompass_Preferred

Cortisol sulfate sodium (Cortisol 21-sulfate) is a metabolite of Cortisol. It is a specific ligand for intracellular transcortin.

  • Purity of 99.79%
  • For research use only
  • Appears as a white to off-white solid
  • Endogenous metabolite
  • Shipped at room temperature in continental US
  • Store at 4°C, sealed, away from moisture
  • Soluble in DMSO at 100 mg/mL
  • Affects behavior through direct action on the central nervous system

Catalog No. 50-004-17425


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.