Learn More
Medchemexpress LLC Cortisone (Standard) | 53-06-5 | 98.51% | 360.44 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY17461R50MG
Encompass_Preferred
Cortisone (Standard) is the analytical standard of Cortisone, intended for research and analytical applications. It is an oxidized metabolite of Cortisol and acts as an immunosuppressant and anti-inflammatory agent. This compound is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Oxidized metabolite of cortisol
  • Acts as an immunosuppressant
  • Acts as an anti-inflammatory agent
  • Suitable for HPLC, GC, and MS experiments
Catalog No. 50-003-86705