Learn More
Medchemexpress LLC Cortistatin-14 | 186901-48-4 | 99.98% | 1720.03 | 1 MG
SDP
Supplier:  Medchemexpress LLC 1MGHYP1932
Encompass_Preferred
Cortistatin-14 is a neuropeptide structurally and functionally similar to somatostatin. It binds to somatostatin receptors (sst1-sst5) and exhibits anticonvulsant, neuroprotective, and anti-inflammatory properties.
  • Binds to somatostatin receptors (sst1-sst5)
  • Exhibits anticonvulsant effects
  • Provides neuroprotective effects
  • Possesses anti-inflammatory properties
  • Reduces release of calcitonin gene-related peptide from peripheral sensory nerve endings
  • Decreases secretion of cytokine IL-1β in mouse peritoneal macrophages
  • Reduces acute neurogenic plasma protein leakage
  • Inhibits mechanical hyperalgesia
  • Regulates pituitary secretion
Catalog No. 50-003-65891