Learn More
CPC Scientific Glp-Glu-Arg-Pro-Pro-Leu-Gln-Gln-Pro-Pro-His-Arg-Asp-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CORT003B
Encompass
ONE-LETTER SEQUENCE: Glp-ERPPLQQPPHRDMOLECULAR FORMULA: C68H105N23O21MOLECULAR WEIGHT:1580.7STORAGE CONDITIONS: -20 5CRESEARCH AREA: NeuropeptidesREFERENCES: L. de Leccea et al., Nature, 381, 242 (1996)
Catalog No. 50-210-9142