Learn More
Cayman Chemical Corynoln 10mg
SDP
Supplier:  Cayman Chemical 3071210 MG
Encompass_Preferred
An isoquinoline alkaloid with diverse biological activities; an inhibitor of acetylcholinesterase (IC50 = 30.6 µM); an inhibitor of BACE1 (IC50s = 33.59 and 89.07 µM for FRET and immobilized enzyme reactive assays, respectively); decreases LPS-induced NF-κB activation and reverses LPS-induced decreases in Nrf2 and HO-1 protein levels in HUVECs; increases the survival rate in a mouse model of LPS- and heat-killed E. coli-induced sepsis at 10, 20, and 40 mg/kg; selectively inhibits proliferation of B16/F10 mouse and A375 human melanoma cells (IC50s = 12.87 and 10.47 µM, respectively) over non-cancerous melanocytes (IC50 = 126.61 µM); reduces tumor growth in a B16/F10 mouse melanoma model at 20 mg/kg
Catalog No. 50-298-8721