Learn More
ATCC COS-7; Kidney Fibroblast; African Green Monkey (Chlorocebus aethiops) Grivet
Supplier:  ATCC CRL1651
Encompass
COS-7 is a cell line exhibiting fibroblast morphology that was derived from the CV-1 cell line (ATCC CCL-70) by transformation with an origin defective mutant of SV40 which codes for wild type T antigen. This is an African green monkey kidney fibroblast-like cell line suitable for transfection by vectors requiring expression of SV40 T antigen and can be used in toxicology research.
Catalog No. 50-238-3468