Learn More
Medchemexpress LLC HY-32015 5mg Medchemexpress, Cot inhibitor-1 CAS:915365-57-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY320155MG
Encompass_Preferred
Medchemexpress, HY-32015 5mg Cot inhibitor-1 CAS:915365-57-0 Cot inhibitor-1 (compound 28) is a selective tumor progression loci-2 (Tpl2) kinase inhibitor with an IC50 of 28 nM. Cot inhibitor-1 shows an inhibition of TNF-alpha production in human whole blood with an IC50 of 5.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2227